Ethyl-3-amino-1H-pyrazole-4-carboxylate 99%

Supplier: Thermo Scientific Chemicals

A10882-06 A10882-14 A10882-22
CAAAA10882-06EA 112.97 CAD
CAAAA10882-06 CAAAA10882-14 CAAAA10882-22
Ethyl-3-amino-1H-pyrazole-4-carboxylate 99%
Ethyl 3-amino-1H-pyrazole-4-carboxylate

Purity: 99% CAS No.: 6994-25-8 Molecular Formula: C6H9N3O2 Formula Weight: 155.16 Melting Point: 104-107° Beilstein Reference: 4964

Formula: C₆H₉N₃O₂
Melting Pt: 104…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00005238
CAS Number: 6994-25-8
EINECS: 230-262-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR