Isonicotinoyl chloride hydrochloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Pyridine-4-carbonyl Chloride Hydrochloride, 4-Pyridinoyl chloride hydrochloride
A10863-09
A10863-18
A10863-30
CAAAA10863-09EA
70.63
CAD
CAAAA10863-09
CAAAA10863-18
CAAAA10863-30
Isonicotinoyl chloride hydrochloride 98%
Isonicotinoyl chloride hydrochloride
Purity: 98%
CAS No.: 39178-35-3
Molecular Formula: C6H4ClNO.HCl
Formula Weight: 178.01
Melting Point: 154-162°
Beilstein Reference: 3626052
UN No.: 3261
MDL No.: MFCD00012830
Formula:
C₆H₄ClNO MW: 178.01 g/mol Melting Pt: 154…162 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012830 CAS Number: 39178-35-3 EINECS: 254-331-2 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...