3-Amino-1-phenyl-2-pyrazolin-5-one 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Amino-2,4-dihydro-2-phenyl-3H-pyrazol-3-one
A10862-14
A10862-22
CAAAA10862-14EA
54.67
CAD
CAAAA10862-14
CAAAA10862-22
3-Amino-1-phenyl-2-pyrazolin-5-one 97%
3-Amino-1-phenyl-2-pyrazolin-5-one
Purity: 97%
CAS No.: 4149-06-8
Molecular Formula: C9H9N3O
Formula Weight: 175.19
Melting Point: ca 213° dec.
Beilstein Reference: 153681
MDL No.: MFCD00003137
Formula:
C₉H₉N₃O Melting Pt: ca 213 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00003137 CAS Number: 4149-06-8 EINECS: 223-974-0 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...