4-Bromo-4'-hydroxybiphenyl 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4'-Bromo-[1,1'-biphenyl]-4-ol

A10819-06 A10819-14 A10819-22
CAAAA10819-06EA 92.59 CAD
CAAAA10819-06 CAAAA10819-14 CAAAA10819-22
4-Bromo-4'-hydroxybiphenyl 98%
4-Bromo-4'-hydroxybiphenyl

Purity: 98% CAS No.: 29558-77-8 Molecular Formula: C12H9BrO Formula Weight: 249.11 Melting Point: 163-168° MDL No.: MFCD00059076

Formula: C₁₂H₉BrO
MW: 249.11 g/mol
Melting Pt: 163…168 °C
Storage Temperature: Ambient
MDL Number: MFCD00059076
CAS Number: 29558-77-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR