N-α-Benzoyl-L-arginine 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Bz-Arg-OH, N-α-Benzoyl-L-Arginine

A10812-09 A10812-18
CAAAA10812-09EA 50.88 CAD
CAAAA10812-09 CAAAA10812-18
N-α-Benzoyl-L-arginine 99%
N-alpha-Benzoyl-L-arginine

Purity: 99% CAS No.: 154-92-7 Molecular Formula: C13H18N4O3 Formula Weight: 278.31 Storage Temperature: 2°C to 8°C Melting Point: 285° dec. MDL No.: MFCD00001763

Formula: C₁₃H₁₈N₄O₃
MW: 278.312 g/mol
Melting Pt: 285 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00001763
CAS Number: 154-92-7
EINECS: 205-837-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR