7,7,8,8-Tetracyanoquinodimethane 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10779-06
A10779-14
CAAAAA10779-06EA
172.22
CAD
CAAAAA10779-06
CAAAAA10779-14
7,7,8,8-Tetracyanoquinodimethane 98%
7,7,8,8-Tetracyanoquinodimethane
Fieser: 1,1136 12,464
Purity: 98%
CAS No.: 1518-16-7
Molecular Formula: C12H4N4
Formula Weight: 204.19
Melting Point: ca 288° dec.
Beilstein Reference: 611604
UN No.: 3439
MDL No.: MFCD00011664
Formula:
C₁₂H₄N₄ Melting Pt: ca 288 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00011664 CAS Number: 1518-16-7 EINECS: 216-174-8 UN: 3439 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...