3,4-Pyridinedicarboxylic acid anhydride 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Furo[3,4-c]pyridine-1,3-dione

A10771-03 A10771-06 A10771-14
CAAAA10771-03EA 59.88 CAD
CAAAA10771-03 CAAAA10771-06 CAAAA10771-14
3,4-Pyridinedicarboxylic acid anhydride 97%
3,4-Pyridinedicarboxylic acid anhydride

Purity: 97% CAS No.: 4664-08-8 Molecular Formula: C7H3NO3 Formula Weight: 149.11 Melting Point: 75-79° MDL No.: MFCD00010680

Formula: C₇H₃NO₃
MW: 149.11 g/mol
Melting Pt: 75…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00010680
CAS Number: 4664-08-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR