1-Cyclohexene-1-carboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

A10741-03 A10741-06 A10741-14
CAAAAA10741-03EA 71.73 CAD
CAAAAA10741-03 CAAAAA10741-06 CAAAAA10741-14
1-Cyclohexene-1-carboxylic acid 97%
1-Cyclohexene-1-carboxylic acid

Purity: 97% CAS No.: 636-82-8 Molecular Formula: C7H10O2 Formula Weight: 126.16 Flash Point: 110°(230°F) Refractive Index: 1.4990 Melting Point: 30-34° Boiling Point: 138-140°/14mm Beilstein Reference: 1905520 UN No.: 3261 MDL No.: MFCD00001545

Formula: C₇H₁₀O₂
Boiling Pt: 138…140 °C/14 mm
Melting Pt: 30…34 °C
Density: 1.100
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001545
CAS Number: 636-82-8
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR