3,5-Dichloro-2-hydroxybenzenesulfonic acid sodium salt 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10732-09
A10732-18
A10732-30
CAAAA10732-09EA
90.06
CAD
CAAAA10732-09
CAAAA10732-18
CAAAA10732-30
3,5-Dichloro-2-hydroxybenzenesulfonic acid sodium salt 99%
3,5-Dichloro-2-hydroxybenzenesulphonic acid sodium salt
Purity: 99%
CAS No.: 54970-72-8
Molecular Formula: C6H3Cl2NaO4S
Formula Weight: 265.05
Melting Point:
Formula:
C₆H₃Cl₂NaO₄S Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009798 CAS Number: 54970-72-8 EINECS: 259-416-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...