2-Furoyltrifluoroacetone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4,4,4-Trifluoro-1-(furan-2-yl)butane-1,3-dione, 4,4,4-Trifluoro-1-(2-furyl)-1,3-butanedione

A10693-06 A10693-14
CAAAA10693-06EA 81.84 CAD
CAAAA10693-06 CAAAA10693-14
2-Furoyltrifluoroacetone 98%
2-Furoyltrifluoroacetone

Purity: 98% CAS No.: 326-90-9 Molecular Formula: C8H5F3O3 Formula Weight: 206.12 Flash Point: 87°(188°F) Refractive Index: 1.5280 Melting Point: 20-21° Boiling Point: 203° Beilstein Reference: 168085 MDL No.: MFCD00020935

Formula: C₈H₅F₃O₃
MW: 206.12 g/mol
Boiling Pt: 203 °C
Melting Pt: 20…21 °C
Density: 1.391
Flash Pt: 87 °C (188 °F)
Storage Temperature: Ambient
MDL Number: MFCD00020935
CAS Number: 326-90-9
EINECS: 206-315-1
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR