Benzil dioxime 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Diphenylglyoxime

A10687-14 A10687-22
CAAAA10687-14EA 93.54 CAD
CAAAA10687-14 CAAAA10687-22
Benzil dioxime 98%
Benzil dioxime

Purity: 98% CAS No.: 23873-81-6 Molecular Formula: C14H12N2O2 Formula Weight: 240.26 Melting Point: ca 238° dec. Beilstein Reference: 2053615 MDL No.: MFCD00002113

Formula: C₁₄H₁₂N₂O₂
MW: 240.26 g/mol
Melting Pt: ca 238 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00002113
CAS Number: 23873-81-6
EINECS: 245-921-0
Merck Index: 14,1079

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR