DL-Tartaric acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10683-36
A10683-0B
A10683-30
CAAAAA10683-36EA
159.58
CAD
CAAAAA10683-36
CAAAA10683-0B
CAAAA10683-30
DL-Tartaric acid 99%
DL-Tartaric acid
Purity: 99%
CAS No.: 133-37-9
Molecular Formula: C4H6O6
Formula Weight: 150.09
Melting Point: 210-212° dec.
Beilstein Reference: 1725148
MDL No.: MFCD00071626
Formula:
C₄H₆O₆ Melting Pt: 210…212 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00071626 CAS Number: 133-37-9 EINECS: 205-105-7 Merck Index: 14,9069 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...