Phenylglyoxal monohydrate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,2-Dihydroxy-1-phenylethan-1-one, Benzoylformaldehyde, Phenylglyoxal hydrate, 2,2-Dihydroxyacetophenone
A10677-06
A10677-14
A10677-22
CAAAA10677-06EA
116.6
CAD
CAAAA10677-06
CAAAA10677-14
CAAAA10677-22
Phenylglyoxal monohydrate 97%
Phenylglyoxal monohydrate
Purity: 97%
CAS No.: 1075-06-5
Molecular Formula: C8H6O2.H2O
Formula Weight: 152.15 (134.12anhy)
Melting Point: 71-77°
Boiling Point: 142°/125mm
Beilstein Reference: 1854721
MDL No.: MFCD00149499
Formula:
C₈H₆O₂ MW: 152.15 g/mol (134.12 anhy) Boiling Pt: 142 °C/125 mm Melting Pt: 71…77 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149499 CAS Number: 1075-06-5 EINECS: 214-036-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...