Methyl-3-(methylthio)propionate 98%
Supplier: Thermo Scientific Chemicals
Warning
|
A10649-14
A10649-22
A10649-36
CAAAA10649-14EA
51.82
CAD
CAAAA10649-14
CAAAA10649-22
CAAAA10649-36
Methyl-3-(methylthio)propionate 98%
Methyl 3-(methylthio)propionate
Purity: 98%
CAS No.: 13532-18-8
Molecular Formula: C5H10O2S
Formula Weight: 134.20
Flash Point: 72°(161°F)
Refractive Index: 1.4650
Boiling Point: 72-73°/11mm
Beilstein Reference: 1745077
MDL No.: MFCD00008565
Formula:
C₅H₁₀O₂S Boiling Pt: 72…73 °C/11 mm Density: 1.074 Flash Pt: 72 °C(161 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008565 CAS Number: 13532-18-8 EINECS: 236-883-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...