N-(tert-Butoxycarbonyl)-L-leucine hydrate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Boc-L-Leucine, BOC-L-Leucine hydrate, Boc-Leu-OH hydrate, N-Boc-L-leucine hydrate

A10647-06 A10647-14
CAAAA10647-06EA 37.76 CAD
CAAAA10647-06 CAAAA10647-14
N-(tert-Butoxycarbonyl)-L-leucine hydrate 99%
N-(tert-Butoxycarbonyl)-L-leucine hydrate

Purity: 99% CAS No.: 13139-15-6 Molecular Formula: C11H21NO4.xH2O Formula Weight: 231.29(anhy) Storage Temperature: 2°C to 8°C Melting Point: 84-87° Beilstein Reference: 1711814 Optical Rotation: -25° (c=2 in acetic acid) MDL No.: MFCD00066067

Formula: C₁₁H₂₁NO₄·xH₂O
MW: 231.29 g/mol (anhy)
Melting Pt: 84…87 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00066067
CAS Number: 13139-15-6
EINECS: 236-073-2

Order Now

Specification Test Results

Optical rotation -25° (c=2 in acetic acid)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR