Methyl (diethylphosphono)acetate 97%

Supplier: Thermo Scientific Chemicals
Warning

A10644-14 A10644-22 A10644-36
CAAAA10644-14EA 96.38 CAD
CAAAA10644-14 CAAAA10644-22 CAAAA10644-36
Methyl (diethylphosphono)acetate 97%
Methyl (diethylphosphono)acetate

Purity: 97% CAS No.: 1067-74-9 Molecular Formula: C7H15O5P Formula Weight: 210.17 Flash Point: 110°(230°F) Refractive Index: 1.4320 Boiling Point: 126-128°/8mm Beilstein Reference: 1782397 MDL No.: MFCD00009081

Formula: C₇H₁₅O₅P
Boiling Pt: 126…128 °C/8 mm
Density: 1,145
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009081
CAS Number: 1067-74-9
EINECS: 213-938-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR