Acetaldehyde oxime (mixture of syn and anti) 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10640-18
A10640-30
CAAAA10640-18EA
214.88
CAD
CAAAA10640-18
CAAAA10640-30
Acetaldehyde oxime (mixture of syn and anti) 98%
Acetaldehyde oxime (mixture of syn and anti)
Fieser: 1,3
Purity: 98%
CAS No.: 107-29-9
Molecular Formula: CH3CH=NOH
Formula Weight: 59.07
Flash Point: 36°(96°F)
Refractive Index: 1.4260
Odor: Characteristic
Boiling Point: 115-117°
Beilstein Reference: 1209252
Merck Reference: 14,42
UN No.: 2332
MDL No.: MFCD00002124
Formula:
C₂H₅NO Boiling Pt: 115…117 °C Density: 0.969 Flash Pt: 36 °C(96 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002124 CAS Number: 107-29-9 EINECS: 203-479-6 UN: 2332 ADR: "3",III Merck Index: 14,42 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...