3-Chloropropanesulfonyl chloride ≥98%

Supplier: Thermo Scientific Chemicals
Danger

A10631-09 A10631-18 A10631-30
CAAAA10631-09EA 147.26 CAD
CAAAA10631-09 CAAAA10631-18 CAAAA10631-30
3-Chloropropanesulfonyl chloride ≥98%
3-Chloropropanesulfonyl chloride

Purity: 98+% CAS No.: 1633-82-5 Molecular Formula: C3H6Cl2O2S Formula Weight: 177.05 Flash Point: 110°(230°F) Refractive Index: 1.4900 Boiling Point: 70°/0.5mm Beilstein Reference: 1754119 UN No.: 3265 MDL No.: MFCD00007463

Formula: C₃H₆Cl₂O₂S
Boiling Pt: 70 °C/0.5 mm
Density: 1.456
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007463
CAS Number: 1633-82-5
EINECS: 216-646-3
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR