Bis(cyclohexanone)oxalyldihydrazone 98%

Supplier: Thermo Scientific Chemicals

A10628-14 A10628-22
CAAAA10628-14EA 93.43 CAD
CAAAA10628-14 CAAAA10628-22
Bis(cyclohexanone)oxalyldihydrazone 98%
Bis(cyclohexanone)oxalyldihydrazone

Purity: 98% CAS No.: 370-81-0 Molecular Formula: C14H22N4O2 Formula Weight: 278.36 Melting Point: 208-212° Beilstein Reference: 2388004 MDL No.: MFCD00001659

Formula: C₁₄H₂₂N₄O₂
Melting Pt: 208…212 °C
Storage Temperature: Ambient
MDL Number: MFCD00001659
CAS Number: 370-81-0
EINECS: 206-729-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR