3-Amino-4-bromo-5-phenyl-1H-pyrazole 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10621-03
A10621-06
CAAAA10621-03EA
39.82
CAD
CAAAA10621-03
CAAAA10621-06
3-Amino-4-bromo-5-phenyl-1H-pyrazole 96%
3-Amino-4-bromo-5-phenyl-1H-pyrazole
Purity: 96%
CAS No.: 2845-78-5
Molecular Formula: C9H8BrN3
Formula Weight: 238.09
Odor: Odorless
Melting Point: 111-117°
MDL No.: MFCD00082666
Formula:
C₉H₈BrN₃ Melting Pt: 111…117 °C Storage Temperature: Ambient |
MDL Number:
MFCD00082666 CAS Number: 2845-78-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...