Pivalamidine hydrochloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10613-06
A10613-14
CAAAA10613-03EA
162.95
CAD
CAAAA10613-03
CAAAA10613-06
CAAAA10613-14
Pivalamidine hydrochloride 98%
Pivalamidine hydrochloride
Purity: 98%
CAS No.: 18202-73-8
Molecular Formula: C5H12N2.HCl
Formula Weight: 136.63
Melting Point: 188-193°
Beilstein Reference: 3618865
MDL No.: MFCD00051988
Formula:
C₅H₁₂N₂ Melting Pt: 188…193 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051988 CAS Number: 18202-73-8 EINECS: 242-091-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...