Methyl-4-(bromomethyl)benzoate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Methyl 4-(bromomethyl)benzoate
A10557-22
A10557-06
A10557-14
CAAAAA10557-22EA
843.72
CAD
CAAAAA10557-22
CAAAA10557-06
CAAAA10557-14
Methyl-4-(bromomethyl)benzoate 98%
Methyl 4-(bromomethyl)benzoate
Purity: 98%
CAS No.: 2417-72-3
Molecular Formula: C9H9BrO2
Formula Weight: 229.07
Melting Point: 53-58°
Boiling Point: 130-135°/2mm
UN No.: 1759
MDL No.: MFCD00032453
Formula:
C₉H₉BrO₂ MW: 229.07 g/mol Boiling Pt: 130…135 °C (2 mmHg) Melting Pt: 53…58 °C Storage Temperature: Ambient |
MDL Number:
MFCD00032453 CAS Number: 2417-72-3 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...