Salicylaldehyde azine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10527-09
A10527-18
CAAAA10527-09EA
165.9
CAD
CAAAA10527-09
CAAAA10527-18
Salicylaldehyde azine 97%
Salicylaldehyde azine
Purity: 97%
CAS No.: 959-36-4
Molecular Formula: C14H12N2O2
Formula Weight: 240.26
Melting Point: 216-222°
Beilstein Reference: 1913597
Formula:
C₁₄H₁₂N₂O₂ Melting Pt: 216…222 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043496 CAS Number: 959-36-4 EINECS: 213-500-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...