Benzohydroxamic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10509-14
A10509-30
A10509-09
A10509-18
CAAAAA10509-14EA
278.08
CAD
CAAAAA10509-14
CAAAAA10509-30
CAAAA10509-09
CAAAA10509-18
Benzohydroxamic acid 98%
Benzohydroxamic acid
Purity: 98%
CAS No.: 495-18-1
Molecular Formula: C7H7NO2
Formula Weight: 137.14
Storage Temperature: 2°C to 8°C
Melting Point: ca 125° dec.
Beilstein Reference: 1907585
MDL No.: MFCD00002109
Formula:
C₇H₇NO₂ Melting Pt: ca 125 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00002109 CAS Number: 495-18-1 EINECS: 207-797-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...