1,2-Naphthoquinone-4-sulfonic acid sodium salt 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sodium 3,4-dioxo-3,4-dihydronaphthalene-1-sulfonate, Folin's reagent, Sodium 3,4-dioxo-3,4-dihydronaphthalene-1-sulphonate

A10507-14 A10507-22
CAAAA10507-14EA 175.38 CAD
CAAAA10507-14 CAAAA10507-22
1,2-Naphthoquinone-4-sulfonic acid sodium salt 97%
1,2-Naphthoquinone-4-sulphonic acid sodium salt

Colorimetric determination of amino acids and amines.


Purity: 97% CAS No.: 521-24-4 Molecular Formula: C10H5NaO5S Formula Weight: 260.20 Melting Point: ca 289° dec. Solubility: Soluble in water. Moderately soluble in acetone. Slightly soluble in alcohol Beilstein Reference: 4169788 Merck Reference: 14,8646 MDL No.: MFCD00001700


Soluble in water. Moderately soluble in acetone. Slightly soluble in alcohol

Formula: C₁₀H₅NaO₅S
MW: 260.20 g/mol
Melting Pt: ∼ 289 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00001700
CAS Number: 521-24-4
EINECS: 208-308-9
Merck Index: 14,8646

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR