Sulfonazo III sodium salt ≥96%
Supplier: Thermo Scientific Chemicals
A10486-06
A10486-14
CAAAA10486-06EA
169.06
CAD
CAAAA10486-06
CAAAA10486-14
Sulfonazo III sodium salt ≥96%
Sulfonazo III sodium salt
Spectrophotometric reagent for alkaline earth metals and indicator for sulfate titrations
CAS No.: 68504-35-8
Molecular Formula: C22H12N4Na4O14S4
Formula Weight: 776.57
Melting Point: ≈300°
Beilstein Reference: 1900148
MDL No.: MFCD00003942
Formula:
C₂₂H₁₆N₄O₁₄S₄ Melting Pt: >=300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003942 CAS Number: 68504-35-8 EINECS: 270-924-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...