Diethylammonium diethyldithiocarbamate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Diethyldithiocarbamic Acid Diethylammonium Salt, Diethylammonium diethylcarbamodithioate

A10458-06 A10458-14 A10458-22
CAAAA10458-06EA 78.37 CAD
CAAAA10458-06 CAAAA10458-14 CAAAA10458-22
Diethylammonium diethyldithiocarbamate 98%
Diethylammonium diethyldithiocarbamate

Purity: 98% CAS No.: 1518-58-7 Molecular Formula: C9H22N2S2 Formula Weight: 222.42 Melting Point: 81-84° MDL No.: MFCD00013146

Formula: C₅H₁₁NS₂
Melting Pt: 81…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00013146
CAS Number: 1518-58-7
EINECS: 216-175-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR