Ethyl-4-bromocrotonate tech. 80%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(E)-Ethyl 4-bromobut-2-enoate, Ethyl trans-4-bromo-2-butenoate
A10371-09
A10371-18
A10371-30
CAAAA10371-09EA
59.25
CAD
CAAAA10371-09
CAAAA10371-18
CAAAA10371-30
Ethyl-4-bromocrotonate tech. 80%, Technical Grade
Ethyl 4-bromocrotonate
Purity: 80%
CAS No.: 37746-78-4
Molecular Formula: BrCH2CH=CHCO2CH2CH3
Formula Weight: 193.05
Storage Temperature: 2°C to 8°C
Flash Point: 97°(206°F)
Refractive Index: 1.4930
Boiling Point: 94-95°/12mm
Beilstein Reference: 1721388
UN No.: 1760
MDL No.: MFCD00000247
Formula:
C₆H₉BrO₂ MW: 193.05 g/mol Boiling Pt: 94…95 °C/12 mm Density: 1.402 Flash Pt: 97 °C(206 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000247 CAS Number: 37746-78-4 UN: 1760 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...