(2E)-3-[4-(Acetyloxy)-3-methoxyphenyl]acrylic acid ≥98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(4-Acetoxy-3-methoxyphenyl)acrylic acid, 4-Acetoxy-3-methoxycinnamic acid

A10361-03 A10361-06 A10361-14
CAAAA10361-03EA 45.19 CAD
CAAAA10361-03 CAAAA10361-06 CAAAA10361-14
(2E)-3-[4-(Acetyloxy)-3-methoxyphenyl]acrylic acid ≥98%
(2E)-3-[4-(Acetyloxy)-3-methoxyphenyl]acrylic acid

Purity: 98+% CAS No.: 2596-47-6 Molecular Formula: C12H12O5 Formula Weight: 236.22 Odor: Odorless Melting Point: 193-196° Beilstein Reference: 2218026 MDL No.: MFCD00014378

Formula: C₁₂H₁₂O₅
MW: 236.23 g/mol
Melting Pt: 193…196 °C
Storage Temperature: Ambient
MDL Number: MFCD00014378
CAS Number: 2596-47-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR