3-Butylidenephthalide mixture E/Z 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Butylideneisobenzofuran-1(3H)-one

A10353-09 A10353-18
CAAAA10353-09EA 58.93 CAD
CAAAA10353-09 CAAAA10353-18 CAAAA10353-30
3-Butylidenephthalide mixture E/Z 95%
3-Butylidenephthalide

Purity: 95% CAS No.: 551-08-6 Molecular Formula: C12H12O2 Formula Weight: 188.23 Flash Point: 132°(269°F) Refractive Index: 1.5770 Boiling Point: 108-109°/0.1mm Beilstein Reference: 5145 MDL No.: MFCD00047319

Formula: C₁₂H₁₂O₂
Boiling Pt: 108…109 °C/0.1 mm
Density: 1.103
Flash Pt: 132 °C(269 °F)
Storage Temperature: Ambient
MDL Number: MFCD00047319
CAS Number: 551-08-6
EINECS: 208-991-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR