3-Aminophthalic acid hydrochloride dihydrate 98%

Supplier: Thermo Scientific Chemicals

A10319-03 A10319-06 A10319-14
CAAAA10319-03EA 62.73 CAD
CAAAA10319-03 CAAAA10319-06 CAAAA10319-14
3-Aminophthalic acid hydrochloride dihydrate 98%
3-Aminophthalic acid hydrochloride dihydrate

Purity: 98% CAS No.: 6946-22-1 Molecular Formula: C8H7NO4.HCl.2H2O Formula Weight: 253.65 (217.62anhy) Melting Point: 182° dec. MDL No.: MFCD00150282

Formula: C₈H₇NO₄.HCl.2H₂O
MW: 253.65 g/mol (217.62 anhy)
Melting Pt: 182 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00150282
CAS Number: 6946-22-1
EINECS: 230-106-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR