6-Chlorothiochroman-4-one 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10304-03
A10304-06
CAAAA10304-03EA
48.03
CAD
CAAAA10304-03
CAAAA10304-06
6-Chlorothiochroman-4-one 98%
6-Chlorothiochroman-4-one
Purity: 98%
CAS No.: 13735-12-1
Molecular Formula: C9H7ClOS
Formula Weight: 198.67
Melting Point: 65-69°
Beilstein Reference: 124621
MDL No.: MFCD00052997
Formula:
C₉H₇ClOS Melting Pt: 65…69 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052997 CAS Number: 13735-12-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...