4-(4-Methylphenyl)-4-oxobutyric acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10290-06
A10290-14
CAAAA10290-06EA
86.43
CAD
CAAAA10290-06
CAAAA10290-14
CAAAA10290-22
4-(4-Methylphenyl)-4-oxobutyric acid 98%
4-(4-Methylphenyl)-4-oxobutyric acid
Purity: 98%
CAS No.: 4619-20-9
Molecular Formula: C11H12O3
Formula Weight: 192.22
Melting Point: 124-128°
Beilstein Reference: 2099653
MDL No.: MFCD00020541
Formula:
C₁₁H₁₂O₃ Melting Pt: 124…128 °C Storage Temperature: Ambient |
MDL Number:
MFCD00020541 CAS Number: 4619-20-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...