Atropine sulfate Monohydrate ≥98%

Supplier: Thermo Scientific Chemicals
Danger

A10236-09 A10236-18
CAAAA10236-09EA 114.39 CAD
CAAAA10236-09 CAAAA10236-18
Atropine sulfate Monohydrate ≥98%
Atropine sulfate Monohydrate

Antagonist at muscarinic receptors; also a bronchodilator


Purity: 98+% CAS No.: 5908-99-6 Molecular Formula: C34H46N2O6.H2SO4.H2O Formula Weight: 694.84 (676.82anhy) Melting Point: 187-192° Beilstein Reference: 6109275 UN No.: 1544 MDL No.: MFCD00074815

Formula: C₁₇H₂₃NO₃
MW: 676.82 g/mol
Melting Pt: 187…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00074815
CAS Number: 5908-99-6
EINECS: 200-235-0
UN: 1544
ADR: 6.1 III
Merck Index: 14,875

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR