Methyl-5-acetylsalicylate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10215-14
A10215-22
CAAAA10215-14EA
94.64
CAD
CAAAA10215-14
CAAAA10215-22
Methyl-5-acetylsalicylate 98%
Methyl 5-acetylsalicylate
Purity: 98%
CAS No.: 16475-90-4
Molecular Formula: C10H10O4
Formula Weight: 194.19
Melting Point: 58-62°
Beilstein Reference: 2643824
Formula:
C₁₀H₁₀O₄ Melting Pt: 58…62 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040924 CAS Number: 16475-90-4 EINECS: 240-532-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...