3-Coumaranone 97%
Supplier: Thermo Scientific Chemicals
A10202-03
A10202-06
A10202-14
CAAAA10202-03EA
95.91
CAD
CAAAA10202-03
CAAAA10202-06
CAAAA10202-14
3-Coumaranone 97%
3-Coumaranone
Purity: 97%
CAS No.: 7169-34-8
Molecular Formula: C8H6O2
Formula Weight: 134.13
Storage Temperature: -30°C to -10°C
Melting Point: 97-102°
Beilstein Reference: 115296
MDL No.: MFCD00051810
Formula:
C₈H₆O₂ Melting Pt: 97…102 °C Storage Temperature: Freezer |
MDL Number:
MFCD00051810 CAS Number: 7169-34-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...