(S)-Methyl-2-aminopropanoate hydrochloride 99%

Supplier: Thermo Scientific Chemicals

Synonyms: H-Ala-OMe·HCl, H-Ala-OMe.HCl, L-Alanine methyl ester hydrochloride, (S)-2-Aminopropionic Acid Methyl Ester Hydrochloride

A10178-06 A10178-14 A10178-22
CAAAA10178-06EA 49.3 CAD
CAAAA10178-06 CAAAA10178-14 CAAAA10178-22
(S)-Methyl-2-aminopropanoate hydrochloride 99%
(S)-Methyl-2-aminopropanoate hydrochloride

Purity: 99% CAS No.: 2491-20-5 Molecular Formula: C4H9NO2.HCl Formula Weight: 139.58 Storage Temperature: 2°C to 8°C Melting Point: 106-110° Beilstein Reference: 3594033 Optical Rotation: +6.5° (c=10 in methanol) MDL No.: MFCD00063663

Formula: C4H10ClNO2
Melting Pt: 106…110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063663
CAS Number: 2491-20-5
EINECS: 219-652-4

Order Now

Specification Test Results

Optical rotation +6.5° (c=10 in methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR