tert-Butylacetyl chloride 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A10162-14
A10162-22
A10162-36
CAAAA10162-14EA
112.5
CAD
CAAAA10162-14
CAAAA10162-22
CAAAA10162-36
tert-Butylacetyl chloride 98+%
tert-Butylacetyl chloride
Purity: 98+%
CAS No.: 7065-46-5
Molecular Formula: (CH3)3CCH2COCl
Formula Weight: 134.61
Storage Temperature: 2°C to 8°C
Flash Point: 21°(69°F)
Refractive Index: 1.4230
Odor: Acrid
Boiling Point: 128-130°
Beilstein Reference: 1740421
UN No.: 2924
MDL No.: MFCD00000737
Formula:
C₆H₁₁ClO Boiling Pt: 128…130 °C Density: 0.967 Flash Pt: 21 °C (69 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000737 CAS Number: 7065-46-5 EINECS: 230-356-4 UN: 2924 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...