2-Methyl-3-(2-furyl)propenal 97%

Supplier: Thermo Scientific Chemicals
Warning

A10125-09 A10125-18
CAAAA10125-09EA 80.58 CAD
CAAAA10125-09 CAAAA10125-18 CAAAA10125-30
2-Methyl-3-(2-furyl)propenal 97%
2-Methyl-3-(2-furyl)propenal

Purity: 97% CAS No.: 874-66-8 Molecular Formula: C8H8O2 Formula Weight: 136.15 Storage Temperature: 2°C to 8°C Flash Point: 100°(212°F) Refractive Index: 1.6140 Boiling Point: 280-281° Beilstein Reference: 2438 MDL No.: MFCD00063240

Formula: C₈H₈O₂
Boiling Pt: 280…281 °C
Density: 1.097
Flash Pt: >100 °C(212 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00063240
CAS Number: 874-66-8
EINECS: 212-866-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR