3,4-Methylenedioxycinnamic acid predominantly trans 99%

Supplier: Thermo Scientific Chemicals
Warning

A10122-14 A10122-22 A10122-36
CAAAA10122-14EA 112.18 CAD
CAAAA10122-14 CAAAA10122-22 CAAAA10122-36
3,4-Methylenedioxycinnamic acid predominantly trans 99%
3,4-Methylenedioxycinnamic acid

Purity: 99% CAS No.: 2373-80-0 Molecular Formula: C10H8O4 Formula Weight: 192.17 Melting Point: ca 243° dec. Beilstein Reference: 84599 MDL No.: MFCD00005837

Formula: C₁₀H₈O₄
Melting Pt: ca 243 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005837
CAS Number: 2373-80-0
EINECS: 219-151-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR