Tungsten dichloride dioxide 99%

Supplier: Thermo Scientific Chemicals
Danger

89962-09 89962-18
CAAA89962-09EA 306.52 CAD
CAAA89962-09 CAAA89962-18
Tungsten dichloride dioxide 99%
Tungsten dichloride dioxide

Purity: 99% CAS No.: 13520-76-8 Molecular Formula: WO2Cl2 Formula Weight: 286.75 Physical Form: Powder Sensitivity: Moisture Sensitive UN No.: 3260 MDL No.: MFCD00054136

Formula: Cl₂O₂W
MW: 286.75 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00054136
CAS Number: 13520-76-8
EINECS: 236-862-1
UN: 3260
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR