Tungsten oxide ≥99.99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
89949-18
89949-30
CAAA89949-18EA
161.16
CAD
CAAA89949-18
CAAA89949-30
Tungsten oxide ≥99.99%
Tungsten oxide
Tungsten oxide is used as a pigment in ceramics and paints. It is used in various types of infrared absorbing coatings and foils.
It is involved in the production of electrochromic windows, which is an electrically switchable glass that change light transmission properties with an applied voltage. It is a unique material utilized for various applications due their chromism, photocatalysis, and sensing capabilities.
Formula:
WO2.9 MW: 230.25 g/mol Melting Pt: 1500…1600 °C Density: 7.16 Storage Temperature: Ambient |
MDL Number:
MFCD00011466 CAS Number: 12037-58-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...