Zinc molybdate ≥98%
Supplier: Thermo Scientific Chemicals
89937-06
89937-14
CAAA89937-06EA
101.91
CAD
CAAA89937-06
CAAA89937-14
Zinc molybdate ≥98%
Zinc molybdate
Purity: 98%
CAS No.: 13767-32-3
Molecular Formula: ZnMoO4
Formula Weight: 225.32
Physical Form: Powder
Melting Point:
Formula:
MoO₄Zn Melting Pt: >700 °C Storage Temperature: Ambient |
MDL Number:
MFCD00049622 CAS Number: 13767-32-3 EINECS: 237-377-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...