Oxone®, monopersulfate, Oxone®
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
KMPS, Potassium hydrogen monopersulfate, PMPS, Potassium peroxymonosulphate, Potassium peroxymonosulfate, Potassium hydrogen monopersulphate, Potassium monopersulfate triple salt, Potassium monopersulphate triple salt
Oxone is used for halogenation of a,b-unsaturated carbonyl compounds and catalytic generation of hypervalent iodine reagents for alcohol oxidation. It is a used for rapid, and good synthesis of oxaziridines.
CAS No.: 70693-62-8
Molecular Formula: 2KHSO5.KHSO4.K2SO4
Formula Weight: 615.50
Physical Form: Granular
Density: 1.15
Sensitivity: Hygroscopic
Appearance: White
UN No.: 3084
MDL No.: MFCD00040551
Formula:
2KHSO₅·KHSO₄·K₂SO₄ MW: 615.50 g/mol Density: 1.15 Storage Temperature: Ambient |
MDL Number:
MFCD00040551 CAS Number: 70693-62-8 EINECS: 274-778-7 UN: 3084 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...