Lead(II) acetylacetonate

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Lead(II) 2,4-pentanedionate

89858-09 89858-18
CAAA89858-09EA 176.96 CAD
CAAA89858-09 CAAA89858-18
Lead(II) acetylacetonate
Lead (II) acetylacetonate

CAS No.: 15282-88-9 Molecular Formula: C10H14O4Pb Formula Weight: 405.41 Physical Form: Crystalline Sensitivity: Hygroscopic Odor: Odorless UN No.: 3467 MDL No.: MFCD00013499

Formula: C₁₀H₁₄O₄Pb
MW: 405.41 g/mol
Melting Pt: 141…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00013499
CAS Number: 15282-88-9
EINECS: 239-323-9
UN: 3467
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR