Calcium pyrophosphate ≥96%

Supplier: Thermo Scientific Chemicals
Warning

89836-30 89836-A1 89836-A7
CAAA89836-30EA 59.88 CAD
CAAA89836-30 CAAA89836-A1 CAAA89836-A7
Calcium pyrophosphate ≥96%
Calcium pyrophosphate

Purity: 96% min CAS No.: 7790-76-3 Molecular Formula: Ca2P2O7 Formula Weight: 254.10 Physical Form: Powder Density: 3.09 Appearance: White Odor: Odorless Melting Point: 1230° MDL No.: MFCD00015983

Formula: Ca₂O₇P₂
Melting Pt: 1230 °C
Density: 3,09
Storage Temperature: Ambient
MDL Number: MFCD00015983
CAS Number: 7790-76-3
EINECS: 232-221-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR