Triisopropoxyvanadium(V) oxide ≥96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
89798-03
89798-06
89798-14
89798-22
89798-36
CAAA89798-03EA
55.62
CAD
CAAA89798-03
CAAA89798-06
CAAA89798-14
CAAA89798-22
CAAA89798-36
Triisopropoxyvanadium(V) oxide ≥96%
Triisopropoxyvanadium(V) oxide
Purity: 96%
CAS No.: 5588-84-1
Molecular Formula: C9H21O4V
Formula Weight: 242.20
Flash Point: 45°(113°F)
Physical Form: Liquid
Density: 1.04
Sensitivity: Moisture Sensitive
Appearance: Pale yellow-orange
Melting Point: -12°
Boiling Point: 81-83°/2mm
UN No.: 1993
MDL No.: MFCD00015017
Formula:
C₉H₂₁O₄V Boiling Pt: 81…83 °C (2 mmHg) Melting Pt: –12 °C Density: 1,04 Flash Pt: 45 °C (113 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00015017 CAS Number: 5588-84-1 EINECS: 226-997-4 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...