Gold platinum palladium ≥99.9% (metals basis), powder APS <5 micron
Supplier: Thermo Scientific Chemicals
|
Warning
|
89726-03
89726-06
CAAA89726-03EA
586.18
CAD
CAAA89726-03
CAAA89726-06
Gold platinum palladium ≥99.9% (metals basis), powder APS <5 micron
Gold platinum palladium
Purity: 99.9%
Molecular Formula: Au:Pt:Pd; 40:40:20 wt%
UN No.: 3089
MDL No.: MFCD00192403
Formula:
Au:Pt:Pd; 40:40:20 % Storage Temperature: Ambient |
MDL Number:
MFCD00192403 UN: 3089 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...