Tin(II) tetrafluoroborate 50% (w/w) in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

89669-A1 89669-D5
CAAA89669-A1EA 164.32 CAD
CAAA89669-A1 CAAA89669-D5
Tin(II) tetrafluoroborate 50% (w/w) in aqueous solution
Tin (II) tetrafluoroborate

CAS No.: 13814-97-6 Molecular Formula: B2F8Sn Formula Weight: 292.30 Physical Form: Liquid Density: 1.670 Appearance: Colorless UN No.: 3264 MDL No.: MFCD00054047

Formula: B₂F₈Sn
MW: 292.30 g/mol
Density: 1.670
Storage Temperature: Ambient
MDL Number: MFCD00054047
CAS Number: 13814-97-6
EINECS: 237-487-6
UN: 3264
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR