Titanium(IV) isobutoxide
Supplier: Thermo Scientific Chemicals
|
Danger
|
89455-18
89455-30
89455-A1
CAAA89455-18EA
68.41
CAD
CAAA89455-18
CAAA89455-30
CAAA89455-A1
Titanium(IV) isobutoxide
Titanium(IV) isobutoxide
CAS No.: 681-98-1
Molecular Formula: C16H36O4Ti
Formula Weight: 340.34
Flash Point: 77°(171°F)
Physical Form: Melt
Density: 1.02
Refractive Index: 1.4950
Sensitivity: Moisture Sensitive
Appearance: Yellow
Boiling Point: 296°
MDL No.: MFCD00015045
Formula:
C₁₆H₃₆O₄Ti Boiling Pt: 296 °C Density: 1,02 Flash Pt: 77 °C (171 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00015045 CAS Number: 681-98-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...